missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM28 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16484-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descrizione
Phosphoinositide-binding protein which associates with both cell and endoplasmic reticulum (ER) membranes. Can bind to the ER membrane protein VAPA and recruit VAPA to plasma membrane sites, thus linking these intracellular compartments. The ORP3-VAPA complex stimulates RRAS signaling which in turn attenuates integrin beta-1 (ITGB1) activation at the cell surface. With VAPA, may regulate ER morphology. Has a role in regulation of the actin cytoskeleton, cell polarity and cell adhesion. Binds to phosphoinositides with preference for PI(3,4)P2 and PI(3,4,5)P3. Also binds 25-hydroxycholesterol and cholesterol.Specifica
RBM28 | |
Polyclonal | |
Unconjugated | |
RBM28 | |
RBM28, RNA binding motif protein 28, RNA binding protein 28 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
312182, 55131, 68272 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8CGC6, Q9NW13 | |
RBM28 | |
RBM28 Fusion Protein Ag9606 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |