Learn More
Invitrogen™ Rat SAP102 (aa 83-100) Synthetic Peptide
Synthetic Peptide
Marca: Invitrogen™ PEP023
Detalles adicionales : CAS : 69-74-9 Peso : 0.01000kg
Descripción
Rat SAP102 (residues 83-100) Synthetic Peptide, PEP-023, from Invitrogen.
PEP-023 is an 18 amino acid (amino acids 83-100) synthetic peptide whose sequence is found in rat SAP102. The sequence of this peptide is (amino to carboxy terminus): G83-P-V-P-P-K-P-V-P-G-K-N-T-P-K-L-N-G100This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and SAP102, catalog # PA1-045. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody.Reconstitute with 0.1 ml of distilled water.Especificaciones
58948 | |
purified | |
Rat SAP102 (aa 83-100) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Rat | |
Q62936 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
Dlg3 | |
No preservative | |
50 μg | |
G83 - P - V - P - P - K - P - V - P - G - K - N - T - P - K - L - N - G100 |