Learn More
Novus Biologicals™ Rat MMP-17 ELISA Kit (Colorimetric)
Colorimetric ELISA Kit
Marca:  Novus Biologicals™ NBP3-06890
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18357933
  • 651.17€ / 1 set
Fecha estimada de envío: 25-06-2024
para ver el stock
Especificaciones
0.16 to 10 ng/mL
ELISA Kit (Colorimetric)
This kit recognizes Rat MMP-17 in samples. No significant cross-reactivity or interference between Rat MMP-17 and analogues was observed.
4326
1 Kit
Storage of components varies. See protocol for specific instructions.
Rat
0.09 ng/mL
Cell Culture Supernatant, Plasma, Serum
ELISA, Sandwich ELISA
MMP17
Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cytoskeleton Markers, Signal Transduction
matrix metallopeptidase 17 (membrane-inserted), matrix metalloproteinase 17 (membrane-inserted), matrix metalloproteinase-17, membrane-type matrix metalloproteinase 4, membrane-type-4 matrix metalloproteinase, MMP-17, MT4MMP, MT4-MMP, MT-MMP 4, MTMMP4