missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB34 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 27435-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Protein transport. Involved in the redistribution of lysosomes to the peri-Golgi region. Plays a role in the maturation of phagosomes that engulf pathogens, such as S.aureus and M.tuberculosis. Plays a role in the fusion of phagosomes with lysosomes.Especificaciones
RAB34 | |
Polyclonal | |
Unconjugated | |
RAB34 | |
RAB34, RAB39, RAH, Ras related protein Rab 34, Ras related protein Rab 39, Ras related protein Rah | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
83871 | |
-20°C | |
Liquid |
Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P0DI83 | |
RAB34 | |
RAB34 Fusion Protein Ag26696 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |