Learn More
Ácido (R)-3-(boc-amino)-2-(fmoc-amino)propiónico, 95 %, Thermo Scientific Chemicals
Catalog Number p-7054290
Click to view available options
Cantidad:
250 mg
1 g
5 g
198544-42-2
C23H26N2O6
426.469
MFCD00798648
PKAUMAVONPSDRW-LJQANCHMSA-N
fmoc-d-dap boc-oh, n2-fmoc-n3-boc-d-2,3-diaminopropionic acid, fmoc-n3-boc-d-2,3-diaminopropionic acid, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-tert-butoxycarbonyl amino propanoic acid, fmoc-n-beta-boc-d-alpha,beta-diaminopropionicacid, 2r-3-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, ambotzfaa1320, fmoc-d-dpr boc-oh, fmoc-d-dapa boc-oh, fmoc-dap boc-oh hydrate
17040127
Ácido (2R)-2-(9h-flúor-9-ilmetoxicarbonilamino)-3-[(2-metilpropan-2-il)oxicarbonilamino]propanoico
CC(C)(C)OC(=O)NCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
198544-42-2
C23H26N2O6
426.469
MFCD00798648
PKAUMAVONPSDRW-LJQANCHMSA-N
fmoc-d-dap boc-oh, n2-fmoc-n3-boc-d-2,3-diaminopropionic acid, fmoc-n3-boc-d-2,3-diaminopropionic acid, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-tert-butoxycarbonyl amino propanoic acid, fmoc-n-beta-boc-d-alpha,beta-diaminopropionicacid, 2r-3-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, ambotzfaa1320, fmoc-d-dpr boc-oh, fmoc-d-dapa boc-oh, fmoc-dap boc-oh hydrate
17040127
Ácido (2R)-2-(9h-flúor-9-ilmetoxicarbonilamino)-3-[(2-metilpropan-2-il)oxicarbonilamino]propanoico
CC(C)(C)OC(=O)NCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
(R)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid is used as catalyst or pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
El ácido (R)-3-(boc-amino)-2-(Fmoc-amino) propiónico se utiliza como catalizador o intermedio farmacéutico.
Solubilidad
Insoluble en agua.
Notas
Mantener el recipiente bien cerrado. Almacenar alejado de agentes oxidantes.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material (R)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid
Cantidad 250 mg
Información de solubilidad Insoluble in water.
Formula Weight (peso de la fórmula) 426.46
Porcentaje de pureza 95%
Forma física Sólido