missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTK7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17799-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The function remains unknown.Especificaciones
PTK7 | |
Polyclonal | |
Unconjugated | |
PTK7 | |
CCK 4, CCK4, Colon carcinoma kinase 4, Protein tyrosine kinase 7, PTK7, PTK7 protein tyrosine kinase 7, Tyrosine protein kinase like 7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
301242, 5754, 71461 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.36 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q13308, Q8BKG3 | |
Ptk7 | |
PTK7 Fusion Protein Ag12004 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |