missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTGR2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14164-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
ZADH1 (zinc binding alcohol dehydrogenase, domain containing 1), also known as PTGR2 (prostaglandin reductase 2), PGR2 or 15-oxoprostaglandin 13-reductase, is a 351 amino acid cytoplasmic protein that belongs to the NADP-dependent oxidoreductase L4BD family. Functioning as s a 15-oxoprostaglandin 13-reductase, ZADH1 catalyzes the conversion of 15-keto-prostaglandin E2 to 15-keto-13, 14-dihydro-prostaglandin E2 in a NADPH-dependent manner. ZADH1 overexpression has been found to repress PPAR gamma transcriptional activity and adipocyte differentiation. Widely expressed, ZADH1 is found at highest levels in heart, kidney, liver, pancreas and prostate, with moderate levels found in brain, small intestine, lung, testis and skeletal muscle. ZADH1 exists as a monomer, utilizes NADPH as a cofactor, and undergoes alternative splicing to produce two isoforms that are encoded by a gene located on human chromosome 14q24.3.Especificaciones
PTGR2 | |
Polyclonal | |
Unconjugated | |
PTGR2 | |
PGR2, PRG 2, prostaglandin reductase 2, PTGR2, ZADH1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
145482, 299194, 77219 | |
-20°C | |
Liquid |
Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5BK81, Q8N8N7, Q8VDQ1 | |
Ptgr2 | |
PTGR2 Fusion Protein Ag5352 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |