Learn More
Epredia™ PrintMate™ Embedding Cassettes in Pre-packed Tubes with Lids, aqua
Epredia™ PrintMate Cassettes™ are designed to contain incised tissue safely and securely for tissue processing, embedding and sectioning. Packed in tubes, PrintMate Cassettes are intended for high throughput printing, and provide exceptional scanning accuracy and reliability when used with the Epredia™ PrintMate™ and Epredia™ SlideMate™ barcode scanner.
Marca: Epredia™ A84210107
Detalles adicionales : Peso : 9.00000kg
Descripción
40 Tubes X 75 Cassettes, total 3000 Cassettes with 3000 LidsEspecificaciones
Round Pore | |
Yes | |
Tan | |
Embedding Biopsy Cassette |