Learn More
Epredia™ PrintMate™ Embedding Cassettes with Lids, aqua
Epredia™ PrintMate Cassettes™ are designed to contain incised tissue safely and securely for tissue processing, embedding and sectioning. PrintMate Cassettes provide exceptional scanning accuracy and reliability when used with the Epredia™ PrintMate™ and Epredia™ SlideMate™ barcode scanner.
Marca: Epredia™ A84210071
Detalles adicionales : Peso : 3.00000kg
Especificaciones
Round Pore | |
Yes | |
Blue | |
Embedding Biopsy Cassette |