missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PREP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Especificaciones
Antígeno | PREP |
---|---|
Concentración | 0.13 mg/mL |
Aplicaciones | Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) |
Clasificación | Polyclonal |
Conjugado | Unconjugated |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16818683
|
Proteintech
11536-1-AP-20UL |
20 μL |
140.00€
20 microlitros |
Fecha estimada de envío: 17-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
16808683
|
Proteintech
11536-1-AP-150UL |
150 μL |
399.00€
150 microlitros |
Fecha estimada de envío: 17-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
p33cdk2 associates with cyclin A in human cells. Kinase activity associated with cyclin A-cdc2 is found only in G2-phase. Cdk2 also complexes with cyclins E, D1, and D3. Cyclin E-cdk2 kinase is active in the G1- and S-phases of the cell cycle and is important (as does cyclin A-cdk2) for the progression from G1-to S-phase. The levels of cyclin A-cdk2 is maximal at the G1/S transition and both cdk2 and cyclin A are found associated with DNA in the initiation complex during replication. Rb protein acts as substrate for cdk2-cyclin E and/or cdk2-cyclin A in vivo. Cdk2 is activated and specifically localized to the nucleus during late G1-, S-, and G2-phase.Especificaciones
PREP | |
Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
O70196, P48147, Q9QUR6 | |
19072, 5550, 83471 | |
PREP Fusion Protein Ag2101 | |
Primary | |
-20°C | |
PREP |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
PE, PEP, Post proline cleaving enzyme, PREP, prolyl endopeptidase | |
Prep | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |