missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP2R3C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 24877-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
P2R3C may regulate MCM3AP phosphorylation through phosphatase recruitment. It may play a role in the activation-induced cell death of B-cells.Especificaciones
PPP2R3C | |
Polyclonal | |
Unconjugated | |
PPP2R3C | |
C14orf10, G4 1, G5pr, PPP2R3C | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
362739, 55012, 59032 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q969Q6 | |
Ppp2r3c | |
PPP2R3C Fusion Protein Ag19282 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |