missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1R9A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18801-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Neurabin I is a brain-specific protein which contains an F-actin binding domain, a PDZ domain, a transmembrane-protein-interacting-domain, and a coiled-coil region. As its structure suggests, neurabin I is a promiscuous protein binding to F-actin, protein phosphatase 1, TGN38, and p70 S6 kinase. Found exclusively in brain, this protein is highly concentrated in the synapse and has also been shown to be enriched in the lamellipodia of the growth cone during neuronal development. Neurabin I appears to be a bridging protein by targeting other proteins to the synapse or by linking membrane proteins to the actin cytoskeleton.Especificaciones
PPP1R9A | |
Polyclonal | |
Unconjugated | |
PPP1R9A | |
KIAA1222, Neurabin 1, Neurabin I, NRB1, NRBI, PPP1R9A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
243725, 55607, 84685 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35867, Q9ULJ8 | |
PPP1R9A | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |