Learn More
PPM1H Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA560656
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: EDQASCEVLT VKKKAGAVTS TPNRNSSKRR SSLPNGEGLQ LKENSESEGV SCHYWSLFDG H Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.
PPM1H Polyclonal antibody specifically detects PPM1H in Human samples. It is validated for Immunohistochemistry.Especificaciones
PPM1H | |
Polyclonal | |
Unconjugated | |
PPM1H | |
A430075L18Rik; ARHCL1; C030002B11Rik; KIAA1157; NERPP-2C; neurite extension-related protein phosphatase related to PP2C; Ppm1h; Protein phosphatase 1H; protein phosphatase 1H (PP2C domain containing); protein phosphatase, Mg2+/Mn2+ dependent 1H; protein phosphatase, Mg2+/Mn2+ dependent, 1H; ras homolog gene family, member C like 1; URCC2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57460 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9ULR3 | |
PPM1H | |
Recombinant protein corresponding to Human PPM1H | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only