missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPARG Recombinant Human Protein, Ligand Binding Domain, GST-Tagged
PPARG Recombinant Human Protein, Ligand Binding Domain, GST-Tagged
Marca: Invitrogen PV4546
Detalles adicionales : Peso : 0.50000kg
Descripción
PPARG (peroxisome proliferator activated receptor gamma) ligand binding domain a 59.00 kDa recombinant human protein (amino acids 201-475) expressed as a GST-tagged fusion protein in baculovirus-infected insect cells. PPAR γ-LBD was expressed and purified in the absence of exogenous ligands, making it ideal for ligand binding and coregulator displacement studies.Especificaciones
NP_ 005028 | |
Cell Structure, Pharma and Biopharma, Target and Lead Identification and Validation, Ligand Binding Assay | |
59kDa | |
50μg | |
For research use only. Not for use in diagnostic procedures | |
Peroxisome proliferator activated receptor gamma | |
16 nM PPAR gamma LBD (GST) is the final concentration recommended for the PolarScreen™ PPARγ Competitor Assay, Green (PV6136). 0.5 nM PPAR gamma LBD (GST) is the final concentration recommended for the LanthaScreen™ TR-FRET PPAR gamma Competitive Binding Assay (PV4894) | |
PPARG Recombinant Human Protein | |
Hormone receptors |
0.96mg/mL | |
5468 | |
PPARG Recombinant Human Protein | |
–68 to –85°C | |
1 Each | |
PPARG Recombinant Human Protein | |
aa 201 to 475 | |
Human | |
95% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.