Learn More
POU4F1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA541509
Detalles adicionales : Peso : 0.00010kg
Descripción
Peptide sequence: LEAYFAVQPR PSSEKIAAIA EKLDLKKNVV RVWFCNQRQK QKRMKFSATY Sequence homology: Cow: 92%; Dog: 92%; Guinea Pig: 100%; Horse: 92%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%
POU4F1 Polyclonal antibody specifically detects POU4F1 in Human samples. It is validated for Western Blot.Especificaciones
POU4F1 | |
Polyclonal | |
Unconjugated | |
Pou4f1 | |
Brain-3A; brain-specific homeobox/POU domain protein 3A; Brn3; Brn-3; brn3.0; Brn-3.0; Brn3a; brn-3A; brn3b; E130119J07Rik; etID22526.1; FLJ13449; homeobox/POU domain protein RDC-1; LOW QUALITY PROTEIN: POU domain, class 4, transcription factor 1; Oct-T1; POU class 4 homeobox 1; POU domain, class 4, transcription factor 1; POU4F1; pou4f1 protein; RDC1; RDC-1 | |
Rabbit | |
Protein A | |
RUO | |
5457 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q01851 | |
Pou4f1 | |
Synthetic peptide directed towards the C-terminal of human POU4F1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |