missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonato de potasio sesquihidrato, ACS, 98,5-101,0 %, Thermo Scientific Chemicals
Click to view available options
Cantidad:
100 g
500 g
Descripción
Potassium carbonate sesquihydrate is used for manufacturing of soaps, pigments, printing ink, glass, pottery, other potassium salts and as an analytical lab reagent. It is also useful as a reagent for removing carbon dioxide from ammonia.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Fabricación de jabones, pigmentos, tinta de impresión, vidrio, alfarería, otras sales de potasio, como reactivo analítico de laboratorio
Especificaciones
Nombre del producto químico o material | Potassium carbonate sesquihydrate |
Forma física | cristalino/a/s |
Cantidad | 500 g |
Intervalo de porcentaje de ensayo | 98.5-101.0% |
Índice Merck | 14,7670 |
Información de solubilidad | Soluble in water; Practically insoluble in alcohol |
Formula Weight (peso de la fórmula) | 165.23 (138.21anhydrous) |
Porcentaje de pureza | 98.5 to 101.0% |
Grado | ACS |
Olor | Odorless |
Mostrar más |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'