missing translation for 'onlineSavingsMsg'
Learn More

Bromuro de potasio, ultra seco, 99,98 % (base metálica), Thermo Scientific Chemicals

Catalog Number 11365757
Click to view available options
Cantidad:
25 g
100 g
500 g
Este artículo no se puede devolver. Vea la política de devoluciones
7758-02-3
BrK
119.00
MFCD00011358
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
253877
CHEBI:32030
[K+].[Br-]
Este artículo no se puede devolver. Vea la política de devoluciones
7758-02-3
BrK
119.00
MFCD00011358
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
253877
CHEBI:32030
[K+].[Br-]

Potassium bromide is used in manufacturing of photographic papers, engraving and lithography, in spectroscopy and as an analytical lab reagent. Potassium bromide is employed in antiepileptic medication for dogs. It is utilized in the bromination of alkenes using cerium(IV) ammonium nitrate. It plays an essential role with diacetoxy iodobenzene for bromination of alkenes and alkynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Envasado bajo argón
TRUSTED_SUSTAINABILITY
Punto de ebullición 1435°C
Nombre del producto químico o material Potassium bromide
Punto de fusión 730°C
Nombre de nota Ultra dry
Color Blanco
Forma física Gránulos de malla -10, en ampolla bajo argón
Cantidad 25 g
Notas del porcentaje del ensayo (base metálica)
Índice Merck 14,7618
Información de solubilidad Freely soluble in water. Soluble in alcohol and glycerol. Slightly soluble in diethyl ether.
Nombre IUPAC potassium bromide
Formula Weight (peso de la fórmula) 119.01
Porcentaje de pureza 99.98%
Olor Odorless
Envase Ampolla
Índice de refracción 1.559
Sensibilidad Hygroscopic
Densidad 2.75 g/mL
Mostrar más Mostrar menos

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'