missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromuro de potasio, para espectroscopía, Thermo Scientific Chemicals
Click to view available options
Cantidad:
250 g
1 kg
2.5 kg
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Punto de ebullición | 1435.0°C |
Nombre del producto químico o material | Potassium bromide |
Punto de fusión | 734.0°C |
Cantidad | 2.5 kg |
Fórmula lineal | KBr |
Información de solubilidad | Solubility in water: 650g/L (20°C). Other solubilities: 1020g/L water (100°C), 1.42g/L ethanol (25°C), 217g/L glycerol, 4g/L alcohol |
Nombre IUPAC | potasio;bromuro |
Formula Weight (peso de la fórmula) | 119 |
Porcentaje de pureza | ≥99.0% (Argentometry) |
Grado | Espectroscopia |
Mostrar más |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'