missing translation for 'onlineSavingsMsg'
Learn More

Poli(2-etil-2-oxazolina), P.M. 500000, Thermo Scientific Chemicals

Catalog Number p-7052017
Click to view available options
Cantidad:
100 g
500 g
2.5 kg
Este artículo no se puede devolver. Vea la política de devoluciones
25805-17-8
(C5H9NO)n
99.13
MFCD00134377
NYEZZYQZRQDLEH-UHFFFAOYSA-N
2-ethyl-2-oxazoline, 2-ethyloxazoline, 2-ethyl-4,5-dihydrooxazole, oxazole, 2-ethyl-4,5-dihydro, 2-oxazoline, 2-ethyl, unii-b8cd92t4p5, unii-jm7uh9q3yn, unii-hnx7574gtx, unii-s5dpg6k67i, ethyl oxazoline
66412
2-ethyl-4,5-dihydro-1,3-oxazole
CCC(=O)N(-*)CC-*
Este artículo no se puede devolver. Vea la política de devoluciones
25805-17-8
(C5H9NO)n
99.13
MFCD00134377
NYEZZYQZRQDLEH-UHFFFAOYSA-N
2-ethyl-2-oxazoline, 2-ethyloxazoline, 2-ethyl-4,5-dihydrooxazole, oxazole, 2-ethyl-4,5-dihydro, 2-oxazoline, 2-ethyl, unii-b8cd92t4p5, unii-jm7uh9q3yn, unii-hnx7574gtx, unii-s5dpg6k67i, ethyl oxazoline
66412
2-ethyl-4,5-dihydro-1,3-oxazole
CCC(=O)N(-*)CC-*

Poly(2-ethyl-2-oxazoline) is used as an adhesion promoter in coatings, in heat sealing and remoisten able hot-melt adhesive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Poly (2-ethyl-2-oxazoline)
Nombre de nota M.W. 500,000
Forma física Sólido blanquecino
Información de solubilidad Soluble in water and organic solvents.
Cantidad 100 g

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'