missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POFUT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17764-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Fucose is typically found as a terminal modification of branched chain glycoconjugates, but it also exists in direct O-linkage to serine or threonine residues within cystine knot motifs in epidermal growth factor-like repeats or thrombospondin type-1 repeats. POFUT2 is an O-fucosyltransferase that use THBS type-1 repeats as substrates.Especificaciones
POFUT2 | |
Polyclonal | |
Unconjugated | |
POFUT2 | |
C21orf80, FUT13, KIAA0958, O FucT 2, Peptide O fucosyltransferase 2, POFUT2, protein O fucosyltransferase 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23275, 309686, 80294 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8VHI3, Q9Y2G5 | |
Pofut2 | |
POFUT2 Fusion Protein Ag12148 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |