Learn More
PNMT Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA560355
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: ECWQDKERQL RARVKRVLPI DVHQPQPLGA GSPAPLPADA LVSAFCLEAV SPDLASFQRA LDHITTLLR Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 86%.
The product of this gene catalyzes the last step of the catecholamine biosynthesis pathway, which methylates norepinephrine to form epinephrine (adrenaline). The enzyme also has beta-carboline 2N-methyltransferase activity. This gene is thought to play a key step in regulating epinephrine production.Especificaciones
PNMT | |
Polyclonal | |
Unconjugated | |
Pnmt | |
Noradrenaline N-methyltransferase; PENT; phenylethanolamine N-methylase; phenylethanolamine N-methyltransferase; phenylethanolamine-N-methyltransferase; PNMT; PNMTase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5409 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P11086 | |
Pnmt | |
Recombinant protein corresponding to Human PNMT | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only