missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 55212-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This antibody is specific to PLS1. This antibody is specific to PLS1. It has no cross reaction to PLS3.
Plastins are a family of actin-binding proteins that are conserved throughout eukaryote evolution and expressed in most tissues of higher eukaryotes. In humans, two ubiquitous plastin isoforms (L and T) have been identified. The protein encoded by this gene is a third distinct plastin isoform, which is specifically expressed at high levels in the small intestine. Alternatively spliced transcript variants varying in the 5′ UTR, but encoding the same protein, have been found for this gene. A pseudogene of this gene is found on chromosome 11.Especificaciones
PLS1 | |
Polyclonal | |
Unconjugated | |
PLS1 | |
I plastin, Intestine specific plastin, Plastin 1, plastin 1 (I isoform), PLS1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
102502, 315926, 5357 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q14651, Q3V0K9 | |
Pls1 | |
Peptide | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |