Learn More
PLEKHG4B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA557591
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: YDRVDEEVHR LVLTSNNRLQ QLEHLRELAS LLEGNDQQSC QKGLQLAKEN PQRTEEMVQD FRRGLSAVVS QAECREGELA RWTRSSELCE TVS Highest antigen sequence identity to the following orthologs: Mouse - 26%, Rat - 26%.
PLEKHG4B (Pleckstrin Homology And RhoGEF Domain Containing G4B) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includeRho guanyl-nucleotide exchange factor activity. This gene encodes a large protein that contains a pleckstrin homology domain and may function as a guanine nucleotide exchange factor.Especificaciones
PLEKHG4B | |
Polyclonal | |
Unconjugated | |
PLEKHG4B | |
CTD-2231H16.1; KIAA1909; LOW QUALITY PROTEIN: pleckstrin homology domain-containing family G member 4B; PH domain-containing family G member 4B; pleckstrin homology and RhoGEF domain containing G4B; pleckstrin homology domain containing, family G (with RhoGef domain) member 4B; pleckstrin homology domain-containing family G member 4B; PLEKHG4B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
153478 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96PX9 | |
PLEKHG4B | |
Recombinant protein corresponding to Human PLEKHG4B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only