Learn More
PLEKHG2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA561639
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: PATTPLPEHK SHMVIPAPST AFCPEQGHCA DIHVPTTPAL PKEICSDFTV SVTTPVPKQE GHLDSESPTN IPLTKQGGSR DVQGPDPVCS QPIQPLSWHG SSLDPQGPGD TLPPLPCHLP D Highest antigen sequence identity to the following orthologs: Mouse - 35%, Rat - 34%.
PLEKHG2 may be a transforming oncogene with exchange activity for CDC42. PLEKHG2 may be a guanine-nucleotide exchange factor (GEF) for RAC1 and CDC42.PLEKHG2 activated by the binding to subunits beta and gamma of the heterotrimeric guanine nucleotide-binding protein (G protein).Especificaciones
PLEKHG2 | |
Polyclonal | |
Unconjugated | |
PLEKHG2 | |
AI194308; ARHGEF42; Clg; common site lymphoma/leukemia GEF; Common site lymphoma/leukemia guanine nucleotide exchange factor; common-site lymphoma/leukemia GEF; common-site lymphoma/leukemia guanine nucleotide exchange factor; Cslg; CTB-60E11.4; DKFZp667J2325; FLJ00018; FLJ22458; FLJ38638; LDAMD; LOW QUALITY PROTEIN: pleckstrin homology domain-containing family G member 2; PH domain-containing family G member 2; pleckstrin homology and RhoGEF domain containing G2; pleckstrin homology domain containing, family G (with RhoGef domain) member 2; pleckstrin homology domain-containing family G member 2; Plekhg2; RGD1310112 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64857 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H7P9 | |
PLEKHG2 | |
Recombinant protein corresponding to Human PLEKHG2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only