Learn More
Novus Biologicals™ PLC-delta 3 Recombinant Protein Antigen
Código de producto. 18677996 Tienda Bio Techne Productos
Change view
Click to view available options
:
100 μg
Código de producto.
18677996 100 μg
1 options
Código de producto. 18677996
Marca: Novus Biologicals™ NBP268706PEP
Recombinant Protein
TRUSTED_SUSTAINABILITY
Especificaciones
Para utilizar con (aplicación) Antibody Competition
Formulación PBS and 1 M Urea, pH 7.4.
ID de gen (Entrez) 113026
Peso molecular MolecularWeight-theroretical: 30 kDa
Cantidad 100 μL
Requisitos de almacenamiento Store at -20°C. Avoid freeze/thaw cycles.
Alias de gen EC 3.1.4.11, KIAA1964, MGC71172, Phosphoinositide phospholipase C-delta-3, phospholipase C, delta 3,1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase delta-3, Phospholipase C-delta-3, PLC delta3, PLC-delta-3
Símbolo de gen PLCD3
Propiedades del inhibidor This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-68706. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mL
Categoría de investigación Lipid and Metabolism
Grado de pureza o calidad >80% by SDS-PAGE and Coomassie blue staining
Expresión PLC-delta 3