missing translation for 'onlineSavingsMsg'
Learn More

Plasminogen Ab-1, Mouse Monoclonal Antibody,Lab Vision™ Epredia™

Provide accurate, reproducible results with the Epredia™ Plasminogen Ab-1, Mouse Monoclonal Antibody.

Marca:  Epredia MS-659-P1

Detalles adicionales : Peso : 1.00000kg

 Ver más versiones de este producto

Código de producto. 12601937

  • 432.00€ / 500 microlitros
Fecha estimada de envío: 22-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Plasminogen is a key proenzyme of plasmin in the fibrinolytic and thrombolytic systems, the deficiency of which leads to a mild thrombotic tendency. Plasminogen binds to fibrin as well as to the surface of endothelial cells and monocytes/macrophages, where it is activated to plasmin. Streptokinase is a plasminogen activator widely used in treating blood-clotting disorders. Complexes of streptokinase with human plasminogen can hydrolytically activate other plasminogen molecules to plasmin, which then dissolves blood clots. Apolipoprotein(a) is very similar to plasminogen in terms of amino acid sequence, it inhibits plasmin generation by competing with plasminogen's binding to fibrin and endothelial cells.

Host Species: Mouse

Clone: SBF1

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Human plasminogen protein

Molecular Weight: ∽120kDa

Positive Control: HUVEC cells

Cellular Localization: Secreted protein

Recommended for:

  • Western Blotting (Nonreduced only)

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

Plasminogen Ab-1
Monoclonal
200μg/mL
Mouse
500 μL
Hematopoietic
Human
IgG2a κ
Western Blot
SBF1
Unconjugated
Human plasminogen protein
RUO
Primary
Concentrated
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales