missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-RelB (Ser552) Recombinant Rabbit Monoclonal Antibody (RelBS552-A7), APC, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Thermo Scientific MA537303
Detalles adicionales : Peso : 0.01000kg
Descripción
Phospho-RelB (Ser552) Recombinant Monoclonal antibody specifically detects Phospho-RelB (Ser552) in Human, Mouse samples. It is validated for Flow Cytometry.Especificaciones
Phospho-RelB (Ser552) | |
Monoclonal | |
APC | |
RELB | |
avian reticuloendotheliosis viral (v-rel) oncogene related B; IREL; I-REL; OTTHUMP00000221313; rel b; RELB; REL-B; RELB proto-oncogene, NF-kB subunit; shep; transcription factor RelB; v-rel avian reticuloendotheliosis viral oncogene homolog B; v-rel avian reticuloendotheliosis viral oncogene homolog B (nuclear factor of kappa light polypeptide gene enhancer in B-cells 3); v-rel reticuloen; v-rel reticuloendotheliosis viral oncogene homolog B; v-rel reticuloendotheliosis viral oncogene homolog B, nuclear factor of kappa light polypeptide gene enhancer in B-cells 3 | |
Rabbit | |
Protein A/G | |
RUO | |
19698, 5971 | |
4° C | |
Liquid |
Flow Cytometry | |
RelBS552-A7 | |
PBS with 0.20% BSA and 0.09% sodium azide | |
Q01201, Q04863 | |
RELB | |
A synthetic phospho-peptide corresponding to residues surrounding Ser552 of human phospho RelB. | |
10 Tests | |
Primary | |
Human, Mouse | |
Antibody | |
IgG κ |