missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-p47phox (Ser359) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA538801
Detalles adicionales : Peso : 0.00900kg
Descripción
p47phox (NCF1) is required, along with NCF2 and membrane bound cytochrome b558, for activation of the latent NADPH oxidase. This oxidase is a multicomponent enzyme that is activated to produce superoxide anion. Mutations in the gene can result in granulomatous disease.Especificaciones
Phospho-p47phox (Ser359) | |
Polyclonal | |
Unconjugated | |
NCF1 | |
Neutrophil cytosol factor 1, NCF-1, 47 kDa autosomal chronic granulomatous disease protein, 47 kDa neutrophil oxidase factor, NCF-47K, Neutrophil NADPH oxidase factor 1, Nox organizer 2, Nox-organizing protein 2, SH3 and PX domain-containing protein 1A, p47-phox, NCF1, NOXO2, SH3PXD1A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
653361 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P14598 | |
NCF1 | |
A synthetic phosphopeptide derived from human p47 phox around the phosphorylation site of Ser359 (Q-R-SP-K-P) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |