missing translation for 'onlineSavingsMsg'
Learn More

Phospho-Lck (Tyr505) Rabbit anti-Human, PE, Clone: LckY505-A3, Invitrogen™

Rabbit Monoclonal Antibody

Marca:  Thermo Scientific MA537093

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 17196678

  • 424.00€ / paquete de 100
Fecha estimada de envío: 08-11-2022
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Phospho-Lck (Tyr505) Monoclonal antibody specifically detects Phospho-Lck (Tyr505) in Human samples. It is validated for Flow Cytometry
Especificaciones

Especificaciones

Phospho-Lck (Tyr505)
Monoclonal
PE
LCK
EC 2.7.10.2; Hck-3; IMD22; kinase Lck; Lck; LCK proto-oncogene, Src family tyrosine kinase; Lck1; Lcktkr; Leukocyte C-terminal Src kinase; LSK; Lskt; Lsk-t; Lymphocyte cell-specific protein-tyrosine kinase; lymphocyte protein tyrosine kinase; lymphocyte-specific protein tyrosine kinase; OTTHUMP00000008640; OTTHUMP00000008740; OTTHUMP00000008741; p56(LSTRA) protein-tyrosine kinase; p56; p56lck; p56-LCK; pp58lck; Protein YT16; Proto-oncogene Lck; Proto-oncogene tyrosine-protein kinase LCK; RP4-675E8.4; t cell-specific protein-tyrosine kinase; T-lymphocyte specific protein tyrosine kinase p56lck; tyr; tyrosine-protein kinase Lck; YT16
A synthetic phospho-peptide corresponding to residues surrounding Tyr505 of human phospho Lck.
100 Tests
Primary
Human
Antibody
IgG1 κ
Flow Cytometry
LckY505-A3
PBS with 0.20% BSA and 0.09% sodium azide
P06239
Rabbit
Protein A/G
RUO
3932
2-8°C
Liquid
Videos
SDS
Documentos
Certificados
Ofertas especiales

Ofertas especiales