missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-ETV6 (Ser257) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA564721
Detalles adicionales : Peso : 0.01000kg
Descripción
Phospho-ETV6 (Ser257) Polyclonal Antibody detects endogenous levels of ETV6 only when phosphorylated at Ser257.
This gene encodes a core component of the exosome. The mammalian exosome is required for rapid degradation of AU rich element-containing RNAs but not for poly(A) shortening. The association of this protein with the exosome is mediated by protein-protein interactions with ribosomal RNA-processing protein 42 and ribosomal RNA-processing protein 46.Especificaciones
Phospho-ETV6 (Ser257) | |
Polyclonal | |
Unconjugated | |
ETV6 | |
TEL, TEL1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
2120 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P41212 | |
ETV6 | |
A synthesized peptide derived from human Tel (Phospho-Ser257) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |