missing translation for 'onlineSavingsMsg'
Learn More

Phospho-Btk (Tyr551) Rabbit anti-Human, Biotin, Clone: BtkY551-G12, Invitrogen™

Rabbit Monoclonal Antibody

Marca:  Thermo Scientific MA537213

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 17196798

  • 424.00€ / paquete de 100
Fecha estimada de envío: 31-10-2022
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Phospho-Btk (Tyr551) Monoclonal antibody specifically detects Phospho-Btk (Tyr551) in Human samples. It is validated for Flow Cytometry
Especificaciones

Especificaciones

Phospho-Btk (Tyr551)
Monoclonal
Biotin
BTK
agammaglobulinaemia tyrosine kinase; Agammaglobulinemia tyrosine kinase; AGMX1; AI528679; AT; ATK; B-cell progenitor kinase; Bpk; Bruton agammaglobulinemia tyrosine kinase; Bruton tyrosine kinase; Bruton's tyrosine kinase; Btk; dominant-negative kinase-deficient Brutons tyrosine kinase; IMD1; Kinase EMB; MGC126261; MGC126262; OTTHUMP00000023677; PSCTK1; RP1-164F3.2; truncated Bruton agammaglobulinemia tyrosine kinase; tyrosine-protein kinase BTK; Tyrosine-protein kinase BTK (Brutons tyrosine kinase) (Agammaglobulinaemia tyrosine kinase) (ATK) (B cell progenitor kinase) (BPK) (Kinase EMB); tyrosine-protein kinase BTK isoform (lacking exon 13 to 17); tyrosine-protein kinase BTK isoform (lacking exon 14); xid; XLA; X-linked immune deficiency
A synthetic phospho-peptide corresponding to residues surrounding human Btk (Y551).
100 Tests
Primary
Human
Antibody
IgG1 κ
Flow Cytometry
BtkY551-G12
PBS with 0.20% BSA and 0.09% sodium azide
Q06187
Rabbit
Protein A/G
RUO
695
2-8°C
Liquid
Videos
SDS
Documentos
Certificados
Ofertas especiales

Ofertas especiales