missing translation for 'onlineSavingsMsg'
Learn More

Phospho-Akt1 (Ser473) Rabbit anti-Human, Mouse, Clone: AktS473-C7, Invitrogen™

Rabbit Monoclonal Antibody

Marca:  Thermo Scientific MA536903

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 17196488

  • 552.00€ / 200 microlitros
Fecha estimada de envío: 02-11-2022
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Phospho-Akt1 (Ser473) Monoclonal antibody specifically detects Phospho-Akt1 (Ser473) in Human, Mouse samples. It is validated for Flow Cytometry
Especificaciones

Especificaciones

Phospho-Akt1 (Ser473)
Monoclonal
0.5 mg/mL
PBS with 0.10% BSA, 50% glycerol and 0.02% sodium azide
P31749, P31750
Rabbit
Protein A/G
RUO
11651, 207
-20°C
Liquid
Flow Cytometry
AktS473-C7
Unconjugated
AKT1
Akt; Akt kinase; AKT serine/threonine kinase 1; AKT/PKB; AKT1; AKT1 kinase; AKT1m; Akt1m protein; Akt1-PA; Akt1-PB; Akt1-PC; Akt1-PD; Akt1-PE; CAKT; C-AKT; CG4006; CG4006 gene product from transcript CG4006-RE; CG4006-PA; CG4006-PB; CG4006-PC; CG4006-PD; CG4006-PE; CWS6; dakt; D-Akt; dAkt kinase; dAKT/dPKB; dAkt/PKB; DAKT1; DAKT1/PKB; Dmel\CG4006; Dmel_CG4006; DPKB; DRAC-PK; DRAC-PK66; DRAC-PK85; l(3)04226; l(3)89Bq; murine thymoma viral (v-akt) oncogene homolog 1; pAkt; p-Akt; PKB; PKB alpha; PKB beta; PKB gamma; PKB/AKT; PKB/dAKT; PKBalpha; PKB-ALPHA; PKBG; PRKBA; Protein kinase B; protein kinase B alpha; protein kinase B-alpha; proto-oncogene c-Akt; Rac; rac protein kinase alpha; RAC protein kinase alpha RAC-PK alpha; RAC serine/threonine-protein kinase; RAC-ALPHA; RAC-alpha serine/threonine-protein kinase; RacPK; RAC-PK-alpha; RAC-PK-beta; RAC-PK-gamma; related to A and C kinases; related to PKA to PKC protein kinases; related to the A and C kinases; serine/threonine protein kinase; serine-threonine protein kinase; Thymoma viral proto-oncogene; thymoma viral proto-oncogene 1; v-akt murine thymoma viral oncogene homolog 1; v-akt murine thymoma viral oncogene-like protein 1
A synthetic phospho-peptide corresponding to residues surrounding Ser473 of human phospho Akt1.
200 μL
Primary
Mouse, Human
Antibody
IgG1 κ
Videos
SDS
Documentos
Certificados
Ofertas especiales

Ofertas especiales