Learn More
PGM2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA531378
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Chicken (83%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Catalyzes the conversion of the nucleoside breakdown products ribose-1-phosphate and deoxyribose-1-phosphate to the corresponding 5-phosphopentoses. May also catalyze the interconversion of glucose-1-phosphate and glucose-6-phosphate. Has low glucose 1,6-bisphosphate synthase activity. [UniProt]Especificaciones
PGM2 | |
Polyclonal | |
Unconjugated | |
PGM2 | |
Glucose phosphomutase 2; MSTP006; PGM 2; PGM2; Phosphodeoxyribomutase; phosphoglucomutase 2; phosphoglucomutase-2; Phosphopentomutase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55276 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.41 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q96G03 | |
PGM2 | |
Recombinant fragment corresponding to a region within amino acids 96 and 437 of Human PGM2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |