missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perforin-1 (Pore Forming Protein) (Apoptosis Marker) Mouse, Clone: PRF1/2468, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 5551-MSM2-P1
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Perforin-1 (Pore Forming Protein) (Apoptosis Marker) | |
Cytoplasmic | |
PRF1/2468 | |
Unconjugated | |
Mouse | |
75kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
5551 | |
IgG2b κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant human Perforin-1 protein fragment (apprx. aa 413-552) | |
HDLM-2 Cells. Spleen | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |