missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDS5A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17485-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Nop56p is a yeast nucleolar protein that is part of a complex with the nucleolar proteins Nop58p and fibrillarin. Nop56p is required for assembly of the 60S ribosomal subunit and is involved in pre-rRNA processing. NOL5A is similar in sequence to Nop56p and is also found in the nucleolus. Multiple transcript variants encoding several different isoforms have been found for this gene, but the full-length nature of most of them have not been determined.Nop56p is a yeast nucleolar protein that is part of a complex with the nucleolar proteins Nop58p and fibrillarin. Nop56p is required for assembly of the 60S ribosomal subunit and is involved in pre-rRNA processing. The protein encoded by this gene is similar in sequence to Nop56p and is also found in the nucleolus. Multiple transcript variants encoding several different isoforms have been found for this gene, but the full-length nature of most of them have not been determined.Especificaciones
PDS5A | |
Polyclonal | |
Unconjugated | |
PDS5A | |
KIAA0648, PDS5, PDS5A, PIG54, SCC 112, SCC112 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23244, 305343, 71521 | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A4L9P7, Q29RF7, Q6A026 | |
Pds5a | |
PDS5A Fusion Protein Ag11525 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |