Learn More
Novus Biologicals™ PDK-1 Recombinant Protein Antigen
Código de producto. 18680788 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0,1 mL
Tamaño de la unidad:
0.10 ml
18680788 0,1 mL 0.10 ml
1 options
Código de producto. 18680788 Proveedor Novus Biologicals™ N.º de proveedor NBP248631PEP
Recombinant Protein
TRUSTED_SUSTAINABILITY
Especificaciones
Para utilizar con (aplicación) Antibody Competition
Formulación PBS and 1 M Urea, pH 7.4.
ID de gen (Entrez) 5170
Peso molecular MolecularWeight-theroretical: 26 kDa
Cantidad 0,1 mL
Requisitos de almacenamiento Store at -20°C. Avoid freeze/thaw cycles.
Alias de gen 3-phosphoinositide-dependent protein kinase 1, 3-phosphoinositide-dependent protein kinase 2 pseudogene, PDK1, PDPK2, PDPK2P, PkB kinase like gene 1
Símbolo de gen PDPK1
Propiedades del inhibidor This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-48631. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mLFor current lot information, including availability, please contact our technical support team click nb-technical@bio-techne.com For further blocking peptide related information and a protocol, click here .
Categoría de investigación Cancer, mTOR Pathway, Signal Transduction
Grado de pureza o calidad >80% by SDS-PAGE and Coomassie blue staining
Expresión PDK-1