missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGFRB Monoclonal Antibody (18A2), APC, Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen A15719
Detalles adicionales : Peso : 0.00280kg
Descripción
PDGFR (Platelet Derived Growth Factor Receptor) exhibits two different isoforms, alpha and beta coded by two different genes. They possess five immunoglobulin like domains that are involved in ligand binding. Two tyrosine kinase domains are separated by a kinase insert to which PI-3 kinase can bind. PDGF AA can bind autophosphorylate and activate only PDGFR, alpha while PDGF BB can bind and activate to both PDGFR, alpha and beta.Especificaciones
PDGFRB | |
Monoclonal | |
APC | |
PDGFRB | |
PDGF Receptor beta, JTK12, PDGFR, PDGFR1, CD140 antigen-like family member B, PDGF-R-beta, beta-type platelet-derived growth factor receptor | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
5159 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
18A2 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P09619 | |
PDGFRB | |
CD140B-transfected NIH-3T3 cells | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |