missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Parathyroid Hormone Rabbit, Clone: PTH/1717R, enQuireBio™
Rabbit Monoclonal Antibody
Marca: EnquireBio 5741-RBM7-P0
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Parathyroid Hormone | |
Cytoplasmic and Secreted | |
PTH/1717R | |
Unconjugated | |
NP_001303281 | |
Rabbit | |
9kDa | |
Protein A/G Purified | |
RUO | |
5741 | |
IgG κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
PTH | |
Recombinant human PTH protein N-terminal fragment | |
Human parathyroid gland carcinoma | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |