Learn More
p53 Ab-8 Mouse Monoclonal Antibody, Epredia™
Deliver accurate, reproducible results with the Epredia™ p53 Ab-8, Mouse Monoclonal Antibody.
Marca: Epredia MS-738-R7
Detalles adicionales : Peso : 0.01200kg
Descripción
p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to mdm2, SV40 T antigen and human papilloma virus E6 protein p53 senses DNA damage and possibly facilitating repair. Mutation involving p53 is found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma.Host Species: Mouse
Clone: DO-7 + BP53-12
Isotype: IgG2b + IgG2a
Species Reactivity: Human. Others not known.
Epitope: aa 37-45 and aa 20-25
Immunogen: Recombinant human p53 protein
Molecular Weight: 53kDa
Positive Control: MDA-231, SKBR-3 cells or Colon CA
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Native and denatured)
Especificaciones
p53 Ab-8 | |
Monoclonal | |
— | |
Mouse | |
7 mL | |
Cancer and Tumor Biology | |
Human | |
IgG2a, IgG2b |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
DO-7 + BP53-12 | |
Unconjugated | |
Recombinant human p53 protein | |
IVD | |
Primary | |
Prediluted |