Learn More
p41-ARCb Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA530353
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: A431. Predicted reactivity: Mouse (94%), Rat (93%), Pig (96%), Chicken (80%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes one of seven subunits of the human Arp2/3 protein complex. This subunit is a member of the SOP2 family of proteins and is most similar to the protein encoded by gene ARPC1A. The similarity between these two proteins suggests that they both may function as p41 subunit of the human Arp2/3 complex that has been implicated in the control of actin polymerization in cells. It is possible that the p41 subunit is involved in assembling and maintaining the structure of the Arp2/3 complex. Multiple versions of the p41 subunit may adapt the functions of the complex to different cell types or developmental stages. This protein also has a role in centrosomal homeostasis by being an activator and substrate of the Aurora A kinase.Especificaciones
p41-ARCb | |
Polyclonal | |
Unconjugated | |
ARPC1B | |
41kDa; AA408064; AA408534; AA571392; actin related protein 2/3 complex subunit 1B; actin related protein 2/3 complex subunit 1B, 41kDa; actin related protein 2/3 complex, subunit 1B; actin related protein 2/3 complex, subunit 1B, 41kDa; actin-related protein 2/3 complex subunit 1B; Actin-related protein complex 1b; AF007010; ARC41; Arp2/3 complex 41 kDa subunit; ARP2/3 protein complex subunit p41; Arpc1b; AW208418; L72; OTTHUMP00000206020; p40-ARC; p41-ARC; SOP2Hs | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10095 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.74 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
O15143 | |
ARPC1B | |
Recombinant fragment corresponding to a region within amino acids 155 and 372 of Human p41-ARCb | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |