missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA567746
Detalles adicionales : Peso : 0.01000kg
Descripción
Predicted to react with Mouse and Rat samples.
P2Y10 belongs to the family of G-protein coupled receptors, that are preferentially activated by adenosine and uridine nucleotides. Two alternatively spliced transcript variants encoding the same protein isoform have been found for this gene. P2Y10 has been reported to be expressed in lymphoid cells and in monocytes derived from dendritic cells.Especificaciones
P2Y10 | |
Polyclonal | |
Unconjugated | |
P2RY10 | |
5830408N17Rik; G-protein coupled purinergic receptor P2Y10; LYPSR2; P2RY10; p2Y purinoceptor 10; P2Y10; P2Y-like receptor; purinergic receptor P2Y, G-protein coupled 10; purinergic receptor P2Y, G-protein coupled, 10; purinergic receptor P2Y10; putative P2Y purinoceptor 10; RGD1561912; RP3-333E23.1 | |
Rabbit | |
affinity chromatography | |
RUO | |
27334, 317219, 78826 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O00398, Q8BFU7 | |
P2RY10 | |
Synthesized peptide derived from human P2Y10. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |