Learn More
p27 Kip1 Ab-1, Mouse Monoclonal Antibody,Lab Vision™ Epredia™
Achieve accurate, reproducible results with the Epredia™ p27Kip1 Ab-1, Mouse Monoclonal Antibody.
Marca: Epredia MS-256-P
Detalles adicionales : Peso : 0.00250kg
Descripción
p27Kip1, a candidate tumor suppressor gene, functions as a negative regulator of G1 progression and has been proposed to function as a possible mediator of TGF-Β induced G1 arrest.Host Species: Mouse
Clone: DCS-72.F6
Isotype: IgG1/κ
Species Reactivity: Human, Mouse, Rat, and Dog. Does not react with cow. Others not known.
Epitope: aa 83-197
Immunogen: Mouse recombinant p27 protein
Molecular Weight: 27kDa
Positive Control: MAD109, Ls174T cells
Cellular Localization: Nuclear
Recommended for:
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Especificaciones
p27superscriptKip1superscript Ab-1 | |
Monoclonal | |
200μg/mL | |
Mouse | |
1 mL | |
Cancer and Tumor Biology | |
Canine, Human, Mouse, Rat | |
IgG2a κ |
Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot | |
DCS-72.F6 | |
Unconjugated | |
Mouse recombinant p27 protein | |
RUO | |
Primary | |
Concentrated |