missing translation for 'onlineSavingsMsg'
Learn More

p27 Kip1 Ab-1, Mouse Monoclonal Antibody,Lab Vision™ Epredia™

Achieve accurate, reproducible results with the Epredia™ p27Kip1 Ab-1, Mouse Monoclonal Antibody.

Marca:  Epredia MS-256-P0

Detalles adicionales : Peso : 1.00000kg

 Ver más versiones de este producto

Código de producto. 12600717

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

p27Kip1, a candidate tumor suppressor gene, functions as a negative regulator of G1 progression and has been proposed to function as a possible mediator of TGF-Β induced G1 arrest.

Host Species: Mouse

Clone: DCS-72.F6

Isotype: IgG1/κ

Species Reactivity: Human, Mouse, Rat, and Dog. Does not react with cow. Others not known.

Epitope: aa 83-197

Immunogen: Mouse recombinant p27 protein

Molecular Weight: 27kDa

Positive Control: MAD109, Ls174T cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

p27superscriptKip1superscript Ab-1
Monoclonal
200μg/mL
Mouse
100 μL
Cancer and Tumor Biology
Canine, Human, Mouse, Rat
IgG1 κ
Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot
DCS-72.F6
Unconjugated
Mouse recombinant p27 protein
RUO
Primary
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales