missing translation for 'onlineSavingsMsg'
Learn More

p21WAF1Ab-3 Mouse Monoclonal Antibody, Epredia™

Achieve accurate, reproducible results with the Epredia™ p21WAF1 Ab-3, Mouse Monoclonal Antibody.

Marca:  Epredia MS-230-R7

Detalles adicionales : Peso : 0.01200kg

 Ver más versiones de este producto

Código de producto. 12620637

  • 252.00€ / 7 ml
Fecha estimada de envío: 20-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

p21WAF1/Cip1/Sdi1/Pic1 is a tumor suppressor protein. Expression of p21WAF1 is induced by wild type, but not mutant, p53 suppressor protein. The p21WAF1 protein binds to cyclin/CDK complexes and inhibits their kinase activity thereby stopping cell cycle progression. It also binds to PCNA (proliferating cell nuclear antigen) and blocks DNA replication but not the DNA repair process.

Host Species: Mouse

Clone: DCS-60.2

Isotype: IgG2a

Species Reactivity: Human. Does not react with mouse and rat. Others not known

Epitope: Not determined

Immunogen: Full-length human recombinant p21 protein

Molecular Weight: 21kDa

Positive Control: Ls174T or Raji cells, Colon CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not suitable)
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Flow Cytometry (Not suitable)
Especificaciones

Especificaciones

p21WAF1 Ab-3
Monoclonal
Mouse
7 mL
Cancer and Tumor Biology
Human
IgG2a
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
DCS-60.2
Unconjugated
Full length human recombinant p21 protein
IVD
Primary
Prediluted
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales