missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OTOP3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18819-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
OTOP3 belongs to the otopetrin family.Especificaciones
OTOP3 | |
Polyclonal | |
Unconjugated | |
OTOP3 | |
OTOP3, otopetrin 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
347741, 69602 | |
-20°C | |
Liquid |
Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q7RTS5, Q80UF9 | |
OTOP3 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |