missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Osteopontin Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 22952-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Osteopontin is an arginine-glycine-aspartic acid (RGD)-containing glycoprotein that interacts with integrins and CD44 as major receptors. It is a multifunctional protein involved in bone mineralization, cell adhesion, cell migration, chronic inflammatory disease and transformation. Proteolytic cleavage by thrombin and matrix metalloproteinases close to the integrin-binding Arg-Gly-Asp sequence modulates the function of OPN and its integrin binding properties. Thrombin-cleaved fragments of Osteopontin are overexpressed in maligt glial tumors and provide a molecular niche with survival advantage and provide a novel substrate for plasmin and cathepsin D.Especificaciones
Osteopontin | |
Polyclonal | |
Unconjugated | |
SPP1 | |
BNSP, Bone sialoprotein 1, BSPI, ETA 1, Nephropontin, OPN, Osteopontin, secreted phosphoprotein 1, SPP 1, SPP1, Urinary stone protein, Uropontin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25353, 6696 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.36 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P08721, P10451 | |
SPP1 | |
Osteopontin Fusion Protein Ag19216 | |
150 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |