missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OLFM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11997-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Involved in transforming growth factor beta (TGF-beta)-induced smooth muscle differentiation. TGF-beta induces expression and nuclear translocation of OLFM2 where it binds to SRF, causing its dissociation from the transcriptional repressor HEY2/HERP1 and facilitating binding of SRF to target genes. Plays a role in AMPAR complex organization.Especificaciones
OLFM2 | |
Polyclonal | |
Unconjugated | |
OLFM2 | |
NOE2, Noelin 2, NOELIN2, NOELIN2_V1, olfactomedin 2, OlfC, OLFM2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
244723, 313783, 93145 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O95897, Q568Y7, Q8BM13 | |
Olfm2 | |
OLFM2 Fusion Protein Ag2642 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |