Learn More
OLFM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10079-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This antibody is specific for isoform 3 and isoform 4 (AMY) of Olfm1 with native molecular weights of 54 kDa and 16 kDa respectively. AMY may have an essential role in nerve tissue.
The Olfactomedin family comprises a diverse group of secreted glycoproteins, which includes OLFM1 (Noelin-1), OLFM2 (Noelin-2), OLFM3 (Noelin-3), OLFM4 (Noelin-4), tiarin, pancortin, gliomedin and mycocilin. These proteins are implicated in the development of the nervous system. Specifically, OLFM1 and OLFM2 expression is observed in the neural plate and neural crest, as well as in the cranial ganglia in mouse at E8-10, and later in brain tissue and in the zone of polarizing activity in the limb. Overexpression of OLFM1 causes an excess of neural crest emigrations and prolonged neural crest production. OLFM2 participates in the regulation of the development of the anterior nervous system. An Arg144Gln mutation in OLFM2 has been implicated as a possible cause for open-angle glaucoma (OAG).Especificaciones
OLFM1 | |
Polyclonal | |
Unconjugated | |
OLFM1 | |
AMY, NOE1, NOEL1, Noelin, NOELIN1, OlfA, olfactomedin 1, OLFM1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10439, 56177, 93667 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O88998, Q62609, Q99784 | |
Olfm1 | |
OLFM1 Fusion Protein Ag0125 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |