missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCT-2 (POU2F2) (B-Cell Marker) Mouse, Clone: OCT2/2136, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 5452-MSM1-P0
Detalles adicionales : Peso : 0.01000kg
Especificaciones
OCT-2 (POU2F2) (B-Cell Marker) | |
Nuclear | |
OCT2/2136 | |
Unconjugated | |
Mouse | |
60kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
5452 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant fragment of human OCT2 protein (apprx. aa 112-297) | |
Raji or U-937 cells. Spleen or Tonsil. | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |