missing translation for 'onlineSavingsMsg'
Learn More

Zeiss™ Objetivo Plan Apo S

Código de producto. p-8005565
Click to view available options
Potencia de aumento:
0.5X
1.0X
1.5X
2.3X
Distancia de enfoque:
10 mm
30 mm
60 mm
63 mm
134 mm
Unit Size:
cada uno
This item is not returnable. View return policy

Product Code. 11835133

Brand: Zeiss™ 435224

Este artículo requiere una actualización de precios antes de ser solicitado.

Solicite el precio de este artículo aquí.

This item is not returnable. View return policy

Diseñado para su uso con microscopios Zeiss

Ideal para una gran variedad de aplicaciones

Specifications

Tipo Objetivo Plan Apo S
Distancia de enfoque 10 mm
Potencia de aumento 2.3X
Cantidad 1 unidad
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.